Anti-RYR2 Rabbit Polyclonal Antibody (Biotinylated)

Supplier: AFG BIOSCIENCE LLC
A45962-50UG A45962-100UG
77710-359EA 225 USD
77710-359 77710-358
Anti-RYR2 Rabbit Polyclonal Antibody (Biotinylated)
Antibodies
Type:
Antigen: RYR2
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR