Anti-RARRES1 Rabbit Polyclonal Antibody

Supplier: AFG BIOSCIENCE LLC
A45899-50UL A45899-100UL
77710-247EA 225 USD
77710-247 77710-246
Anti-RARRES1 Rabbit Polyclonal Antibody
Antibodies
Type:
Antigen: RARRES1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR