Anti-CP Rabbit Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A45731-50UG A45731-100UG
77709-957EA 228.38 USD
77709-957 77709-956
Anti-CP Rabbit Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: CP
Clonality: Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR