Anti-AKAP8 Rabbit Polyclonal Antibody

Supplier: AFG BIOSCIENCE LLC
A45717-50UL A45717-100UL
77709-932EA 225 USD
77709-932 77709-931
Anti-AKAP8 Rabbit Polyclonal Antibody
Antibodies
ELISA:1:2000-1:5000, WB:1:500-1:2000, IHC:1:25-1:100

Type:
Antigen: AKAP8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR