Anti-RAB38 Rabbit Polyclonal Antibody

Supplier: AFG BIOSCIENCE LLC
A45593-100UL A45593-50UL
77709-710EA 330 USD
77709-710 77709-711
Anti-RAB38 Rabbit Polyclonal Antibody
Antibodies
Type:
Antigen: RAB38
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR