Anti-GRIN2A Rabbit Polyclonal Antibody (FITC)

Supplier: AFG BIOSCIENCE LLC
A45555-100UG A45555-50UG
77709-646EA 330 USD
77709-646 77709-647
Anti-GRIN2A Rabbit Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: GRIN2A
Clonality: Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR