Anti-ERAS Rabbit Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A45509-100UG A45509-50UG
77709-568EA 334.95 USD
77709-568 77709-569
Anti-ERAS Rabbit Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: ERAS
Clonality: Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR