Anti-ERAS Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A45507-100UG A45507-50UG
77709-564EA 334.95 USD
77709-564 77709-565
Anti-ERAS Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: WB:1:1000-1:5000

Type:
Antigen: ERAS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR