Anti-MAP1S Rabbit Polyclonal Antibody (Biotinylated)

Supplier: AFG BIOSCIENCE LLC
A45362-50UG A45362-100UG
77709-311EA 225 USD
77709-311 77709-310
Anti-MAP1S Rabbit Polyclonal Antibody (Biotinylated)
Antibodies
Type:
Antigen: MAP1S
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR