Anti-ZMAT2 Rabbit Polyclonal Antibody (Biotinylated)

Supplier: AFG BIOSCIENCE LLC
A45352-50UG A45352-100UG
77709-291EA 225 USD
77709-291 77709-290
Anti-ZMAT2 Rabbit Polyclonal Antibody (Biotinylated)
Antibodies
Type:
Antigen: ZMAT2
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR