Anti-RAB3C Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A64304-50UG A64304-100UG
77723-776EA 228.38 USD
77723-776 77723-775
Anti-RAB3C Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: WB:1:1000-1:5000

Type:
Antigen: RAB3C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR