Anti-GJB5 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A48340-50UL A48340-100UL
77714-504EA 228.38 USD
77714-504 77714-503
Anti-GJB5 Rabbit Polyclonal Antibody
Antibodies
ELISA:1:2000-1:10000, IHC:1:30-1:150

Type:
Antigen: GJB5
Clonality: Polyclonal
Clone:
Conjucation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR