Anti-HSPD1 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A48256-100UL A48256-50UL
77714-351EA 334.95 USD
77714-351 77714-352
Anti-HSPD1 Rabbit Polyclonal Antibody
Antibodies
ELISA:1:1000-1:5000, IHC:1:50-1:150

Type:
Antigen: HSPD1
Clonality: Polyclonal
Clone:
Conjucation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR