Anti-ABCC2 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A48061-100UL A48061-50UL
77714-002EA 334.95 USD
77714-002 77714-003
Anti-ABCC2 Rabbit Polyclonal Antibody
Antibodies
ELISA:1:2000-1:10000, IHC:1:30-1:150

Type:
Antigen: ABCC2
Clonality: Polyclonal
Clone:
Conjucation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR