Anti-Da-3 Rabbit Polyclonal Antibody (Biotin)

Supplier: AFG Bioscience
A48045-50UG A48045-100UG
77713-971EA 228.38 USD
77713-971 77713-970
Anti-Da-3 Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type:
Antigen: Da-3
Clonality: Polyclonal
Clone:
Conjucation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Trifolium repens
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR