Anti-Da-3 Rabbit Polyclonal Antibody (FITC)

Supplier: AFG BIOSCIENCE LLC
A48044-100UG A48044-50UG
77713-968EA 334.95 USD
77713-968 77713-969
Anti-Da-3 Rabbit Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: Da-3
Clonality: Polyclonal
Clone:
Conjucation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Trifolium repens
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR