Anti-FADD Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A48020-50UL A48020-100UL
77713-931EA 228.38 USD
77713-931 77713-930
Anti-FADD Rabbit Polyclonal Antibody
Antibodies
ELISA:1:1000-1:5000, IHC:1:50-1:150

Type:
Antigen: FADD
Clonality: Polyclonal
Clone:
Conjucation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR