Anti-UCP1 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A47926-50UL A47926-100UL
77713-779EA 228.38 USD
77713-779 77713-778
Anti-UCP1 Rabbit Polyclonal Antibody
Antibodies
ELISA:1:2000-1:10000, IHC:1:30-1:150

Type:
Antigen: UCP1
Clonality: Polyclonal
Clone:
Conjucation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR