Anti-BMP4 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A47834-50UL A47834-100UL
77713-629EA 228.38 USD
77713-629 77713-628
Anti-BMP4 Rabbit Polyclonal Antibody
Antibodies
ELISA:1:2000-1:5000, WB:1:500-1:2000, IHC:1:30-1:150

Type:
Antigen: BMP4
Clonality: Polyclonal
Clone:
Conjucation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR