Anti-COX18 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A47525-50UL A47525-100UL
77713-102EA 228.38 USD
77713-102 77713-101
Anti-COX18 Rabbit Polyclonal Antibody
Antibodies
ELISA:1:1000-1:2000, IHC:1:25-1:100

Type:
Antigen: COX18
Clonality: Polyclonal
Clone:
Conjucation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR