Anti-CST2 Rabbit Polyclonal Antibody

Supplier: AFG BIOSCIENCE LLC
A47380-50UL A47380-100UL
77712-852EA 225 USD
77712-852 77712-851
Anti-CST2 Rabbit Polyclonal Antibody
Antibodies
ELISA:1:2000-1:5000, WB:1:200-1:1000, IHC:1:50-1:200

Type:
Antigen: CST2
Clonality: Polyclonal
Clone:
Conjucation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR