SM68 Standard 2: Ag, Ge, Hf, Mo, Nb, Sb, Si, Sn, Ta, Ti, W, Zr @ 100 µg/mL in 5% HNO3, tr. HF, VHG, LGC Standards
Supplier: LGC Standards
VHG-SM68-2-500
VHG-SM68-2-100
77697-864EA
357.95
USD
77697-864
77697-863
SM68 Standard 2: Ag, Ge, Hf, Mo, Nb, Sb, Si, Sn, Ta, Ti, W, Zr @ 100 µg/mL in 5% HNO3, tr. HF, VHG, LGC Standards
Standards
Cationic Standards
Multi-Element Standards