Anti-TXNDC5 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A46241-50UL A46241-100UL
77710-847EA 228.38 USD
77710-847 77710-846
Anti-TXNDC5 Rabbit Polyclonal Antibody
Antibodies
ELISA:1:2000-1:10000, WB:1:1000-1:5000, IHC:1:30-1:150

Type:
Antigen: TXNDC5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR