Anti-SORT1 Rabbit Polyclonal Antibody

Supplier: AFG BIOSCIENCE LLC
A47328-50UL A47328-100UL
77712-760EA 228.38 USD
77712-760 77712-759
Anti-SORT1 Rabbit Polyclonal Antibody
Antibodies
Type:
Antigen: SORT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR