Anti-SDHA Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A46830-100UL A46830-50UL
77711-904EA 334.95 USD
77711-904 77711-905
Anti-SDHA Rabbit Polyclonal Antibody
Antibodies
ELISA:1:1000-1:2000, WB:1:200-1:1000, IHC:1:30-1:150

Type:
Antigen: SDHA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR