Anti-rho Rabbit Polyclonal Antibody (Biotin)

Supplier: AFG Bioscience
A46454-50UG A46454-100UG
77711-228EA 228.38 USD
77711-228 77711-227
Anti-rho Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type:
Antigen: rho
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: E. coli
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR