Anti-KRT17 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A47045-50UL A47045-100UL
77712-280EA 228.38 USD
77712-280 77712-279
Anti-KRT17 Rabbit Polyclonal Antibody
Antibodies
ELISA:1:1000-1:5000, IHC:1:30-1:150

Type:
Antigen: KRT17
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR