Anti-GLS Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A46828-50UL A46828-100UL
77711-901EA 228.38 USD
77711-901 77711-900
Anti-GLS Rabbit Polyclonal Antibody
Antibodies
ELISA:1:1000-1:2000, IHC:1:25-1:100

Type:
Antigen: GLS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR