Anti-KDM5C Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A60403-100UL A60403-50UL
77716-457EA 334.95 USD
77716-457 77716-458
Anti-KDM5C Rabbit Polyclonal Antibody
Antibodies
ELISA:1:2000-1:10000, IHC:1:30-1:150

Type:
Antigen: KDM5C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR