Anti-BDH1 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A60361-100UL A60361-50UL
77716-384EA 334.95 USD
77716-384 77716-385
Anti-BDH1 Rabbit Polyclonal Antibody
Antibodies
ELISA:1:1000-1:5000, IHC:1:50-1:200

Type:
Antigen: BDH1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR