Anti-RAB8B Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A60153-100UL A60153-50UL
77716-043EA 334.95 USD
77716-043 77716-044
Anti-RAB8B Rabbit Polyclonal Antibody
Antibodies
ELISA:1:2000-1:5000, IHC:1:30-1:150

Type:
Antigen: RAB8B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR