Anti-COPS5 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A60044-100UL A60044-50UL
77715-863EA 334.95 USD
77715-863 77715-864
Anti-COPS5 Rabbit Polyclonal Antibody
Antibodies
ELISA:1:2000-1:10000, WB:1:1000-1:5000, IHC:1:150-1:500

Type:
Antigen: COPS5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR