Anti-EFNA1 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A49037-50UL A49037-100UL
77715-702EA 228.38 USD
77715-702 77715-701
Anti-EFNA1 Rabbit Polyclonal Antibody
Antibodies
ELISA:1:1000-1:5000, IHC:1:50-1:150

Type:
Antigen: EFNA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR