Anti-LEP Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A49008-50UL A49008-100UL
77715-652EA 228.38 USD
77715-652 77715-651
Anti-LEP Rabbit Polyclonal Antibody
Antibodies
ELISA:1:2000-1:10000, IHC:1:30-1:150

Type:
Antigen: LEP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR