Anti-TEX47 Rabbit Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A65152-50UG A65152-100UG
77725-376EA 228.38 USD
77725-376 77725-375
Anti-TEX47 Rabbit Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: TEX47
Clonality: Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR