Anti-NUP37 Rabbit Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A64579-100UG A64579-50UG
77724-291EA 334.95 USD
77724-291 77724-292
Anti-NUP37 Rabbit Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: NUP37
Clonality: Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR