Anti-CCDC82 Rabbit Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A64510-100UG A64510-50UG
77724-163EA 334.95 USD
77724-163 77724-164
Anti-CCDC82 Rabbit Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: CCDC82
Clonality: Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR