Anti-ANXA7 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A44759-50UL A44759-100UL
77708-219EA 228.38 USD
77708-219 77708-218
Anti-ANXA7 Rabbit Polyclonal Antibody
Antibodies
ELISA:1:1000-1:8000, IHC:1:150-1:300

Type:
Antigen: ANXA7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR