Anti-MLN Rabbit Polyclonal Antibody (Biotinylated)

Supplier: AFG BIOSCIENCE LLC
A44385-100UG A44385-50UG
77707-564EA 334.95 USD
77707-564 77707-565
Anti-MLN Rabbit Polyclonal Antibody (Biotinylated)
Antibodies
Type:
Antigen: MLN
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR