Anti-MLN Rabbit Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A44384-100UG A44384-50UG
77707-562EA 334.95 USD
77707-562 77707-563
Anti-MLN Rabbit Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: MLN
Clonality: Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR