Anti-MLN Rabbit Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A44383-50UG A44383-100UG
77707-561EA 228.38 USD
77707-561 77707-560
Anti-MLN Rabbit Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: MLN
Clonality: Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR