Anti-MAPKAPK5 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A44215-50UL A44215-100UL
77707-267EA 228.38 USD
77707-267 77707-266
Anti-MAPKAPK5 Rabbit Polyclonal Antibody
Antibodies
ELISA:1:2000-1:10000, IHC:1:30-1:150

Type:
Antigen: MAPKAPK5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR