Anti-VLDLR Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A62403-100UL A62403-50UL
77720-187EA 334.95 USD
77720-187 77720-188
Anti-VLDLR Rabbit Polyclonal Antibody
Antibodies
ELISA:1:2000-1:10000, IHC:1:30-1:150

Type:
Antigen: VLDLR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR