Anti-SERPINA12 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A62290-50UL A62290-100UL
77720-003EA 228.38 USD
77720-003 77720-002
Anti-SERPINA12 Rabbit Polyclonal Antibody
Antibodies
ELISA:1:2000-1:10000, IHC:1:30-1:150

Type:
Antigen: SERPINA12
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR