Anti-CISD2 Rabbit Polyclonal Antibody (Biotinylated)

Supplier: AFG BIOSCIENCE LLC
A63366-50UG A63366-100UG
77721-998EA 228.38 USD
77721-998 77721-997
Anti-CISD2 Rabbit Polyclonal Antibody (Biotinylated)
Antibodies
Type:
Antigen: CISD2
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR