Anti-CISD2 Rabbit Polyclonal Antibody (FITC)

Supplier: AFG BIOSCIENCE LLC
A63365-50UG A63365-100UG
77721-996EA 228.38 USD
77721-996 77721-995
Anti-CISD2 Rabbit Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: CISD2
Clonality: Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR