Multi-Element 23 Components US EPA Metals Standard: 100 µg/mL each of Ag,Al,As,Ba,Be,Ca,Cd,Co,Cr,Cu,Fe,K,Mg,Mn,Mo,Na,Ni,Pb,Sb,Se,Tl,V,Zn in 5% HNO3,tr. Tartaric Acid,tr. HF,VHG,LGC Standards
Supplier: LGC Standards
VHG-SM23-100
VHG-SM23-500
77695-556EA
304.65
USD
77695-556
77695-557
Multi-Element 23 Components US EPA Metals Standard: 100 µg/mL each of Ag,Al,As,Ba,Be,Ca,Cd,Co,Cr,Cu,Fe,K,Mg,Mn,Mo,Na,Ni,Pb,Sb,Se,Tl,V,Zn in 5% HNO3,tr. Tartaric Acid,tr. HF,VHG,LGC Standards
Standards
Cationic Standards
Multi-Element Standards