Multi-Element Standard 22 components: 100 µg/g each of Ag, Al, B, Ba, Ca, Cd, Cr, Cu, Fe, Mg, Mn, Mo, Na, Ni, P, Pb, Si, Sn, Ti, V, Zn in Hydrocarbon Oil, VHG, LGC Standards
Supplier: LGC Standards
VHG-V21+K-100-400G VHG-V21+K-100-100G VHG-V21+K-100-200G
77693-311EA 376.85 USD
77693-311 77695-651 77695-652
Multi-Element Standard 22 components: 100 µg/g each of Ag, Al, B, Ba, Ca, Cd, Cr, Cu, Fe, Mg, Mn, Mo, Na, Ni, P, Pb, Si, Sn, Ti, V, Zn in Hydrocarbon Oil, VHG, LGC Standards
Standards Cationic Standards Multi-Element Standards
New Product
Order Now
SPECIFICATIONS
Environmentally Preferable