Multi-Element Standard 22 components: 30 µg/g each of Ag, Al, B, Ba, Ca, Cd, Cr, Cu, Fe, Mg, Mn, Mo, Na, Ni, P, Pb, Si, Sn, Ti, V, Zn in Hydrocarbon Oil, VHG, LGC Standards
Supplier: LGC Standards
VHG-V21+K-30-100G VHG-V21+K-30-200G
77695-657EA 176.15 USD
77695-657 77695-658
Multi-Element Standard 22 components: 30 µg/g each of Ag, Al, B, Ba, Ca, Cd, Cr, Cu, Fe, Mg, Mn, Mo, Na, Ni, P, Pb, Si, Sn, Ti, V, Zn in Hydrocarbon Oil, VHG, LGC Standards
Standards Cationic Standards Multi-Element Standards