PinPort™ for Catheters and Tubing
Supplier: INSTECH LABORATORIES, INC
PNP3F22R-50 PNP3F25-50 PNP3F25R-50 PNP3M PNP3M-50 PNP3F22-50
77679-348BX 404.19 USD
77679-348 77679-350 77679-352 77679-354 77679-356 77679-346
PinPort™ for Catheters and Tubing
Fittings and Connectors
The PinPort™ provides fast, aseptic access to externalized rodent catheters or tubing for dosing or blood sampling. It can reduce study costs and improve compliance with the 3Rs.
  • Fast access
  • Closed system for improved patency
  • Needle free
  • Low cost
  • Manufactured in USA
  • Sterilized by ETO gas
The system consists of a lightweight port and a mating injector which can access the port hundreds of times.
Quickly infuse or withdraw from a catheter in a fraction of the time it takes with plugs and blunt needles. The PinPort™ is a closed system that permits aseptic technique. This can reduce infection, improve patency, and ultimately reduce the number of animals required for a study. To ensure safety, the injector is recessed and provides complete needle stick protection for personnel.
Precaución: For research use only.